Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
kr 0,00
Order Entry
Beta Cyclodextrin, Reference Standard
1154569.
:
USP
:
Beta Cyclodextrin, Reference Standard
1154569.
:
USP
:
kr 5 075,00
1 * 500 mg
-
+
Pack type:
Vial
Pk:
500 mg
Standards:
USP reference standard
CAS No.:
7585-39-9
Description:
Beta Cyclodextrin
Description:
Beta Cyclodextrin