Bis((R)-butyramide), Pharmaceutical Analytical Impurities
1A00540.
: USP
:
- Pack type:Vial
- Pk:10 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:1867585-44-1
- Description:Bis((R)-butyramide)
- Synonym(s):(R)-2-amino-N-((R)-1-amino-1-oxobutan-2-yl)butanamide
- Description:Bis((R)-butyramide)
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.