Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
kr 0,00
Order Entry
Calcium Citrate Malate, Reference Standard
1086425.
:
USP
:
Calcium Citrate Malate, Reference Standard
1086425.
:
USP
:
Price
kr 4 550,00
Log in
for availability
Unit of Measure
1 * 200 mg
-
+
Add to cart
Specifications
Pack type:
Vial
Pk:
200 mg
Standards:
USP reference standard
CAS No.:
142606-53-9
Description:
Calcium Citrate Malate
Description:
Calcium Citrate Malate
Specifications
About this item