Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
kr 0,00
Order Entry
Home
Standards
Organic Standards
Dermatan Sulfate, Reference Standard
Dermatan Sulfate, Reference Standard
Dermatan Sulfate, Reference Standard
1171455.
:
USP
:
Dermatan Sulfate, Reference Standard
1171455.
:
USP
:
kr 15 450,00
1 * 25 mg
-
+
Pack type:
Vial
Pk:
25 mg
Standards:
USP reference standard
CAS No.:
54328-33-5
Description:
Dermatan Sulfate
Description:
Dermatan Sulfate