Norway
Order Entry
Desonide 21-Acetate, Pharmaceutical Analytical Impurities
  1A01420.
 :  USP
 :  
Desonide 21-Acetate, Pharmaceutical Analytical Impurities
  1A01420.
 :  USP
 :  
 
kr 9 525,00
   
  1 * 25 mg

 

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • CAS No.:
    25092-25-5
  • Description:
    Desonide 21-Acetate
  • Synonym(s):
    2-((6aR,6bS,7S,8aS,8bS,11aR,12aS,12bS)-7-Hydroxy-6a,8a,10,10-tetramethyl-4-oxo-1,2,4,6a,6b,7,8,8a,11a,12,12a,12b-dodecahydro-8bH-naphtho[2',1':4,5]indeno[1,2-d][1,3]dioxol-8b-yl)-2-oxoethyl acetate
  • Description:
    Desonide 21-Acetate

 

 

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.