Skip to Header
Skip to Main Content
Skip to Footer
/
Norway
Products
Services
Digital Solutions
Applications
Support
About
kr 0,00
Order Entry
Dibasic Potassium Phosphate, Reference Standard
1551128.
:
USP
:
Dibasic Potassium Phosphate, Reference Standard
1551128.
:
USP
:
: 1551128
: USPH1551128
kr 4 450,00
1 * 500 mg
-
+
Pack type:
Vial
Pk:
500 mg
Standards:
USP reference standard
CAS No.:
7758-11-4
Description:
Dibasic Potassium Phosphate
Description:
Dibasic Potassium Phosphate