Norway
Order Entry
Escitalopram Open Ring, Pharmaceutical Analytical Impurities
Escitalopram Open Ring, Pharmaceutical Analytical Impurities
  1A01760.
 :  USP
 :  
Escitalopram Open Ring, Pharmaceutical Analytical Impurities
  1A01760.
 :  USP
 :  
 
kr 7 000,00
   
  1 * 25 mg

Specifications

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • CAS No.:
    103146-25-4
  • Description:
    Escitalopram Open Ring
  • UN No.:
    3077
  • Synonym(s):
    4-[4-(Dimethylamino)-1-(4-fluorophenyl)-1-hydroxybutyl]-3-(hydroxymethyl)benzonitrile
  • Description:
    Escitalopram Open Ring

Specifications

About this item

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.