Fexofenadine, Pharmaceutical Analytical Impurities
: USP
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.
- CAS No.:1422515-52-3
- Synonym(s):1-(4-(4-(2-carboxypropan-2-yl)phenyl)-4-hydroxybutyl)-4-(hydroxydiphenylmethyl)piperidine 1-oxide
- Description:Fexofenadine N-oxide
- StandardsPack typePk
- Fexofenadine N-oxideFexofenadine N-oxide1A01920.1A01920Pharmaceutical Analytical ImpuritiesVial25 mg1 * 25 mgkr 14 400,00: 1A01920: USPH1A01920
- Fexofenadine OlefinFexofenadine Olefin1A00731.1A00731Pharmaceutical Analytical ImpuritiesVial25 mg1 * 25 mgkr 14 400,00: 1A00731: USPH1A00731