Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
kr 0,00
Order Entry
Home
Standards
Organic Standards
Hydroxyethyl Cellulose, Reference Standard
Hydroxyethyl Cellulose, Reference Standard
Hydroxyethyl Cellulose, Reference Standard
1327088.
:
USP
:
Hydroxyethyl Cellulose, Reference Standard
1327088.
:
USP
:
kr 4 175,00
1 * 60 mg
-
+
Pack type:
Vial
Pk:
60 mg
Standards:
USP reference standard
CAS No.:
9004-62-0
Description:
Hydroxyethyl Cellulose
Description:
Hydroxyethyl Cellulose