/
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Oleic Acid, Reference Standard
1478130.
:
USP
:
Oleic Acid, Reference Standard
1478130.
:
USP
:
kr 4 850,00
1 * 1 g
-
+
Pack type:
Ampule
Pk:
1 g
Standards:
USP reference standard
CAS No.:
112-80-1
Description:
Oleic Acid
Description:
Oleic Acid