Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
kr 0,00
Order Entry
Oxyquinoline Sulfate, Reference Standard
1490103.
:
USP
:
Oxyquinoline Sulfate, Reference Standard
1490103.
:
USP
:
kr 4 700,00
1 * 200 mg
-
+
Pack type:
Vial
Pk:
200 mg
Standards:
USP reference standard
CAS No.:
134-31-6
Description:
Oxyquinoline Sulfate
Description:
Oxyquinoline Sulfate
About this item