Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
kr 0,00
Order Entry
Home
Standards
Organic Standards
Peanut Oil, Reference Standard
Peanut Oil, Reference Standard
Peanut Oil, Reference Standard
Catalog # 1500557.
Supplier:
USP
CAS Number:
Expand image
Peanut Oil, Reference Standard
Catalog # 1500557.
Supplier:
USP
CAS Number:
Price
kr 4 850,00
Log in
for availability
Unit of Measure
1 * 1 g
-
+
Add to cart
Specifications
Pack type:
Ampule
Pk:
1 g
Standards:
USP reference standard
CAS No.:
8002-03-7
Description:
Peanut Oil
Description:
Peanut Oil
Specifications
About this item
Product Details
Product Details