Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Order Entry
Home
Standards
Organic Standards
Pepsin, Reference Standard
Pepsin, Reference Standard
Pepsin, Reference Standard
1510051.
:
USP
:
Pepsin, Reference Standard
1510051.
:
USP
:
kr 4 100,00
1 * 5 Items
-
+
Pack type:
Vial
Pk:
5
Standards:
USP reference standard
CAS No.:
9001-75-6
Description:
Pepsin for Assay
Description:
Pepsin
Pk Info:
5 x 1 g