Tolyloyl Propionamide, Pharmaceutical Analytical Impurities
1A01630.
: USP
:
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:402470-91-1
- Description:Tolyloyl Propionamide
- Synonym(s):N,N-Dimethyl-4-oxo-4-(p-tolyl)butanamide
- Description:Tolyloyl Propionamide
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.