Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
kr 0,00
Order Entry
Glyceryl Monocaprylate, Reference Standard
1295764.
:
USP
:
Glyceryl Monocaprylate, Reference Standard
Catalog # 1295764.
Supplier:
USP
CAS Number:
Price
kr 4 700,00
Log in
for availability
Unit of Measure
1 * 300 mg
-
+
Specifications
Pack type:
Vial
Pk:
300 mg
Standards:
USP reference standard
CAS No.:
26402-26-6
Description:
Glyceryl Monocaprylate
Description:
Glyceryl Monocaprylate
Specifications