VWR®, Test Tubes with Rim
Supplier: VWR Collection
Certificates
About this item
Test tubes made from borosilicate 5.1 glass, neutral glass.
- Thin walled
- Excellent chemical resistance
- Relatively resistant to rapid temperature changes and local heating
The economical alternative for a wide range of uses.
Certifications: DIN 12395.
Product Information
- Catalog #LengthExt.ØMaterialWall thicknessPkAvailabilityPrice
- 212-075370 mm8 mmBorosilicate glass 5.10,4 - 0,5 mm100Log in for availability1 * 100 Itemskr 360,00Specifications:
Specifications
Environmentally Preferable - 212-075975 mm10 mmBorosilicate glass 5.10,4 - 0,5 mm100Log in for availability1 * 100 Itemskr 375,00Specifications:
Specifications
Environmentally Preferable - 212-076075 mm12 mmBorosilicate glass 5.10,4 - 0,5 mm100Log in for availability1 * 100 Itemskr 395,00Specifications:
Specifications
Environmentally Preferable - 212-0754100 mm10 mmBorosilicate glass 5.10,4 - 0,5 mm100Log in for availability1 * 100 Itemskr 390,00Specifications:
Specifications
Environmentally Preferable - 212-0755100 mm12 mmBorosilicate glass 5.10,4 - 0,5 mm100Log in for availability1 * 100 Itemskr 415,00Specifications:
Specifications
Environmentally Preferable - 212-0761130 mm14 mmBorosilicate glass 5.10,4 - 0,5 mm100Log in for availability1 * 100 Itemskr 500,00Specifications:
Specifications
Environmentally Preferable - 212-0762130 mm16 mmBorosilicate glass 5.10,5 - 0,6 mm100Log in for availability1 * 100 Itemskr 570,00Specifications:
Specifications
Environmentally Preferable - 212-0763150 mm25 mmBorosilicate glass 5.10,6 - 0,7 mm50Log in for availability1 * 50 Itemskr 530,00Specifications:
Specifications
Environmentally Preferable - 212-0756160 mm16 mmBorosilicate glass 5.10,5 - 0,6 mm100Log in for availability1 * 100 Itemskr 585,00Specifications:
Specifications
Environmentally Preferable - 212-0757180 mm18 mmBorosilicate glass 5.10,5 - 0,6 mm100Log in for availability1 * 100 Itemskr 690,00Specifications:
Specifications
Environmentally Preferable - 212-0758180 mm20 mmBorosilicate glass 5.10,5 - 0,6 mm100Log in for availability1 * 100 Itemskr 805,00Specifications:
Specifications
Environmentally Preferable - 212-0764200 mm25 mmBorosilicate glass 5.10,6 - 0,7 mm50Log in for availability1 * 50 Itemskr 610,00Specifications:
Specifications
Environmentally Preferable - 212-0765200 mm30 mmBorosilicate glass 5.10,7 - 0,8 mm50Log in for availability1 * 50 Itemskr 1 150,00Specifications:
Specifications
Environmentally Preferable