VWR®, Test Tubes without Rim, Thin Walled
About this item
Test tubes made from borosilicate 7.1 glass, ideal for use as disposable.
- Thin walled
- Good chemical and physical properties
- Not suitable for application related to strong heat or rapid temperature changes
An economically proven alternative in reasonable big packs.
Product Information
- Catalog #LengthExt.ØMaterialWall thicknessPkAvailabilityPrice
- 212-074475 mm10 mmBorosilicate glass0,5 - 0,6 mm1.0001 * 1.000 Itemskr 1 325,00
Specifications
Environmentally Preferable - 212-074575 mm12 mmBorosilicate glass0,5 - 0,6 mm1.0001 * 1.000 Itemskr 1 350,00
Specifications
Environmentally Preferable - 100 mm12 mmBorosilicate glass0,5 - 0,6 mm1.0001 * 1.000 Itemskr 1 450,00
Specifications
Environmentally Preferable - 212-0747100 mm13 mmBorosilicate glass0,5 - 0,6 mm1.0001 * 1.000 Itemskr 1 525,00
Specifications
Environmentally Preferable - 212-0748100 mm14 mmBorosilicate glass0,5 - 0,6 mm1.0001 * 1.000 Itemskr 1 650,00
Specifications
Environmentally Preferable - 100 mm16 mmBorosilicate glass0,6 - 0,7 mm1.0001 * 1.000 Itemskr 1 850,00
Specifications
Environmentally Preferable - 125 mm16 mmBorosilicate glass0,6 - 0,7 mm1.0001 * 1.000 Itemskr 2 150,00
Specifications
Environmentally Preferable - 150 mm16 mmBorosilicate glass0,6 - 0,7 mm1.0001 * 1.000 Itemskr 2 250,00
Specifications
Environmentally Preferable - 160 mm16 mmBorosilicate glass0,6 - 0,7 mm1.0001 * 1.000 Itemskr 2 325,00
Specifications
Environmentally Preferable