24 clear not treated reservoir, sterile, HTS Transwell®
Catalog # 734-1558
Supplier: Corning
- Sterile:+
- Description:HTS Transwell® non treated reservoir
- Seq:07
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Pk:48
For use as reservoir tray with HTS Transwell permeable supports.
- Not treated for cell attachment
- Same footprint as standard 24 well plate but with single rectangular well