MICROFLEX® 94-242 Nitrile Static Dissipative Gloves
Supplier: Ansell
Certificates
About this item
Ansell's first disposable static dissipative gloves reduce risk of fire, explosion, or damage caused by electrostatic discharge.
- Static dissipative glove offers exceptional protection from electrostatic discharge
- Proprietary nitrile formulation for a low vertical resistance below 108 Ω
- TNT™ Chemical Splash Resistance Technology for soft, durable protection against a wide range of chemicals
The MICROFLEX® 94-242 gloves reduce the risk of electrostatic discharge, which can cause spontaneous combustion in atmospheric explosive environments. The gloves provide capacitive and resistive touchscreen capability when working with industrial electronics. Black color provides contrast against potentially combustible dust, while hiding the appearance of oils, dirt, and stains.
Certifications: Arrette du 05/08/2020, ASTM D6319, EC 1935/2004, EC 2023/2006, EC 1935/2004, EC 2023/2006, EN 16350:2014, EN ISO 374-5:2016, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, FDA21 CFR 177-2600-US Food Contact Approved, Personal Protective Equipment Regulation (EU) 2016/425 Category III risks.
Specifications
- Catalog No:
- 112-6423
- 112-6422
- 112-6421
- 112-6420
- 112-6424
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Cuff length:
- Standard
- Standard
- Standard
- Standard
- Standard
- Cuff thickness:
- 0,08 mm (3,1 mil)
- 0,08 mm (3,1 mil)
- 0,08 mm (3,1 mil)
- 0,08 mm (3,1 mil)
- 0,08 mm (3,1 mil)
- Finger thickness:
- 0,16 mm (6,2 mil)
- 0,16 mm (6,2 mil)
- 0,16 mm (6,2 mil)
- 0,16 mm (6,2 mil)
- 0,16 mm (6,2 mil)
- Palm thickness:
- 0,12 mm (4,7 mil)
- 0,12 mm (4,7 mil)
- 0,12 mm (4,7 mil)
- 0,12 mm (4,7 mil)
- 0,12 mm (4,7 mil)
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Anti-static:
- +
- +
- +
- +
- +
- Food contact approved:
- +
- +
- +
- +
- +
Specifications
Product Information
- Catalog #Supplier #Glove sizeColourLengthPkAvailabilityPrice
- Supplier Article Number: 94-242/6.5-7Local Article Number: ANSE94-242/6.5-7Specifications:
More
Specifications
Ambidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeFinishTextured fingersCuff styleBeadedCuff lengthStandardCuff thickness0,08 mm (3,1 mil)Finger thickness0,16 mm (6,2 mil)Palm thickness0,12 mm (4,7 mil)SterilityNon sterileAnti-static+Food contact approved+ - Supplier Article Number: 94-242/7.5-8Local Article Number: ANSE94-242/7.5-8Specifications:
More
Specifications
Ambidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeFinishTextured fingersCuff styleBeadedCuff lengthStandardCuff thickness0,08 mm (3,1 mil)Finger thickness0,16 mm (6,2 mil)Palm thickness0,12 mm (4,7 mil)SterilityNon sterileAnti-static+Food contact approved+ - Supplier Article Number: 94-242/8.5-9Local Article Number: ANSE94-242/8.5-9Specifications:
More
Specifications
Ambidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeFinishTextured fingersCuff styleBeadedCuff lengthStandardCuff thickness0,08 mm (3,1 mil)Finger thickness0,16 mm (6,2 mil)Palm thickness0,12 mm (4,7 mil)SterilityNon sterileAnti-static+Food contact approved+ - Supplier Article Number: 94-242/9.5-10Local Article Number: ANSE94-242/9.5-10Specifications:
More
Specifications
Ambidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeFinishTextured fingersCuff styleBeadedCuff lengthStandardCuff thickness0,08 mm (3,1 mil)Finger thickness0,16 mm (6,2 mil)Palm thickness0,12 mm (4,7 mil)SterilityNon sterileAnti-static+Food contact approved+ - 112-642494-242/10.5-1110,5 - 11Black245 mm (9,6")100Log in for availability1 * 100 Itemskr 455,00Supplier Article Number: 94-242/10.5-11Local Article Number: ANSE94-242/10.5-11Specifications:
More
Specifications
Ambidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeFinishTextured fingersCuff styleBeadedCuff lengthStandardCuff thickness0,08 mm (3,1 mil)Finger thickness0,16 mm (6,2 mil)Palm thickness0,12 mm (4,7 mil)SterilityNon sterileAnti-static+Food contact approved+