Anti-BCAS1/NABC1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-11462R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:BCAS1/NABC1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:542-584/584
- Form:Liquid
- Antigen synonyms:Breast carcinoma amplied sequence 1|BCAS 1|Amplied and overexpressed in breast cancer|Breast carcinoma-amplied sequence 1|AIBC 1|BCAS1_HUMAN|AIBC1|BCAS1|Novel amplied in breast cancer 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:62 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human BCAS1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
NaBC1 is a protein found amplified in most breast carcinoma forms. It is expressed primarily as a cytoplasmic, detergent-stable homodimer that has a tendency to interact with DYNLL1 (PIN) and DYNLL2. Breast tumour lines that exhibit 20q13.2 gene amplification express much higher levels of the protein as compared to the levels found in other breast cancer lines that do not overexpress the NaBC1 mRNA. However, this upregulation does not affect growth rate or anchoring abilities of a cell, indicating the oncogenic properties of NaBC1 differ from that of other oncogenes.
Type: Primary
Antigen: BCAS1/NABC1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 542-584/584
Host: Rabbit
Isotype: IgG
Reactivity: