Norway
Order Entry
PEEK Tubing
  18-1112-53
 :  Cytiva
undefined
PEEK Tubing
  18-1112-53
 :  Cytiva
 : 18-1112-53
 : GEHE18-1112-53
kr 990,00
  1 * 1 Items
  • Description:
    PEEK tubing 1/16", inner Ø 0,75 mm, green
  • Seq:
    44
  • Pk:
    1
Polyetheretherketone tubing (PEEK) is intended for use with AKTA systems.
Green color for easy identification.