Norway
Order Entry
Battery Test Chambers with Safety Packages for Aging, Performance or Stress Tests
Battery Test Chambers with Safety Packages for Aging, Performance or Stress Tests
Supplier:  Binder
Certificates
Battery Test Chambers with Safety Packages for Aging, Performance or Stress Tests
Supplier:  Binder
Package P for performance tests. Hazard classification EUCAR hazard level 4/5/6 (Leakage, loss of mass > 50%, Fire or flame and Rupture respectively)
Product Information
  • Catalog #
    Supplier #
    For
    Pk
    Availability
    Price
  • Package P performance tests
    Package P performance tests
    BIND9418-0585
    9418-0585
    MK 115 climatic chamber
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 9418-0585
    Local Article Number: BIND9418-0585
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Package P performance tests
    Package P performance tests
    BIND9418-0607
    9418-0607
    MK 240 climatic chamber
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 9418-0607
    Local Article Number: BIND9418-0607
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Package P performance tests
    Package P performance tests
    BIND9418-0515
    9418-0515
    MK 720 climatic chamber
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 9418-0515
    Local Article Number: BIND9418-0515
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Package P performance tests
    Package P performance tests
    BIND9422-0071
    9422-0071
    MK 1020 climatic chamber
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 9422-0071
    Local Article Number: BIND9422-0071
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Package P performance tests
    Package P performance tests
    BIND9419-0144
    9419-0144
    MK 56 climatic chamber
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 9419-0144
    Local Article Number: BIND9419-0144
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Package P performance tests
    Package P performance tests
    BIND9422-0288
    9422-0288
    MK 400 climatic chamber
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 9422-0288
    Local Article Number: BIND9422-0288
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
Package P Plus for extended performance tests. Hazard classification EUCAR hazard level 5/6 (Fire or flame and Rupture respectively)
Product Information
  • Catalog #
    Supplier #
    For
    Pk
    Availability
    Price
  • Package P Plus extended performance tests
    Package P Plus extended performance tests
    BIND9423-0076
    9423-0076
    MK 400 climatic chamber
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 9423-0076
    Local Article Number: BIND9423-0076
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Package P Plus extended performance tests
    Package P Plus extended performance tests
    BIND9422-0301
    9422-0301
    MK 1020 climatic chamber
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 9422-0301
    Local Article Number: BIND9422-0301
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Package P Plus extended performance tests
    Package P Plus extended performance tests
    BIND9420-0122
    9420-0122
    MK 240 climatic chamber
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 9420-0122
    Local Article Number: BIND9420-0122
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Package P Plus extended performance tests
    Package P Plus extended performance tests
    BIND9420-0341
    9420-0341
    MK 720 climatic chamber
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 9420-0341
    Local Article Number: BIND9420-0341
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
Package S for stress tests. Hazard classification EUCAR hazard level 6 (Rupture)
Product Information
  • Catalog #
    Supplier #
    For
    Pk
    Availability
    Price
  • Package S stress tests
    Package S stress tests
    BIND9422-0169
    9422-0169
    MK 400 climatic chamber
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 9422-0169
    Local Article Number: BIND9422-0169
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Package S stress tests
    Package S stress tests
    BIND9422-0174
    9422-0174
    MK 1020 climatic chamber
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Supplier Article Number: 9422-0174
    Local Article Number: BIND9422-0174
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
Standard MK chambers
Product Information
  • Catalog #
    Seq
    Description
    Shelves supplied (max.)
    W×D×H int.
    W×D×H ext.
    Pk
    Availability
    Price
  • 004
    Dynamic climate chambers , MK 400, 400 V, 50 Hz
    1(6)
    735×810×700 mm
    1115×1400×1710 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    Supplier Article Number: 9020-0406
    Local Article Number: BIND9020-0406
Product Information
  • Catalog #
    Seq
    Description
    Shelves supplied (max.)
    W×D×H int.
    W×D×H ext.
    Pk
    Availability
    Price
  • 003
    Dynamic climate chamber, MK 240, 400 V, 50 Hz
    1(6)
    735×443×700 mm
    1115×925×1710 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    Supplier Article Number: 9020-0376
    Local Article Number: BIND9020-0376
Product Information
  • Catalog #
    Seq
    Description
    Shelves supplied (max.)
    W×D×H int.
    W×D×H ext.
    Pk
    Availability
    Price
  • 006
    Dynamic climate chambers , MK 1020, 400 V, 50 Hz
    1(11)
    1200×810×1020 mm
    1580×1480×2005 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    Supplier Article Number: 9020-0407
    Local Article Number: BIND9020-0407
Product Information
  • Catalog #
    Seq
    Description
    Shelves supplied (max.)
    W×D×H int.
    W×D×H ext.
    Pk
    Availability
    Price
  • 002
    Dynamic climate chamber, MK 115, 400 V, 50 Hz
    1(4)
    600×400×480 mm
    980×865×1725 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    Supplier Article Number: 9020-0375
    Local Article Number: BIND9020-0375
Product Information
  • Catalog #
    Seq
    Description
    Shelves supplied (max.)
    W×D×H int.
    W×D×H ext.
    Pk
    Availability
    Price
  • 005
    Dynamic climate chamber, MK 720, 400 V, 50 Hz
    1(11)
    1200×600×1020 mm
    1580×1140×2005 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    Supplier Article Number: 9020-0377
    Local Article Number: BIND9020-0377
Product Information
  • Catalog #
    Seq
    Description
    Shelves supplied (max.)
    W×D×H int.
    W×D×H ext.
    Pk
    Availability
    Price
  • 001
    Dynamic climate chamber, MK 56, 230 V, 50 Hz
    1(4)
    400×350×420 mm
    720×778×1445 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    Supplier Article Number: 9020-0374
    Local Article Number: BIND9020-0374