Poster, Periodic Table
Leverandør: EDUCATIONSCIENCE COLLECTION P
761-0477EA 0 NOK
761-0477
Poster, Periodic Table
Tabeller
Periodic table, wall poster. New Swedish redesigned wall poster in strong specially treated fabric with sewn channels at the top and bottom. Exciting images of each element and examples of products in our environment that contain the element. The element boxes contain information on the years of discovery and in which country it has taken place and a list of Swedish discoverers. For each substance, there is an indication of atomic number, atomic weight, name, electron configuration, distribution and half-life of radioactive substances. Solid, liquid or gaseous substance and occurrence in nature are shown with color codes.
  • Size 163×133 cm
Leveringsinformasjon: Supplied with information sheets and aluminium suspension strips.