WindPitch, Wind Turbine
Leverandør: EDUCATIONSCIENCE COLLECTION P
761-0158EA 0 NOK
761-0158
WindPitch, Wind Turbine
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
WindPitch is a miniature real-working wind turbine (wind power generator) designed for students to evaluate the pitch (setting angle) of the profiled blades. Learn how the blade pitch and number of blades affect the power output of the wind turbine to maximize the output power. Experimentation also teaches about stalling or reduction of the wind turbine's rotational speed to protect itself under strong wind conditions.
Leveringsinformasjon: WindPitch includes 3 different types of profiled blades and 1 type of Poly-propylene sheet blades; special 3 Phase AC alternator (similar to the one used in real commercial wind turbine); an LED Voltmeter / Music Maker Module to demonstrate the power created from the wind energy in the form of musical sounds and illuminated LED lights. Also included is a complete curriculum on wind energy with easy to follow experiment manual, assembly guide, and background history on the technology and wind power generators.