Sleeve protector, cut resistant, Aracut® Black 14
Leverandør: Honeywell Safety
112-1653EA 255 NOK
DALL4150058
Sleeve protector, cut resistant, Aracut® Black 14
This two-ply Kevlar® knitted sleeve with thumbhole provides cut and puncture resistance.
  • Mitt finish at the fingers helps to provide the cuff with better support
  • Kevlar® fibre offers good resistance to cuts and protection against thermal hazards
  • Double thickness improves the mechanical and thermal performance
  • Dark colour makes it more soilproof, thus ensuring a longer life
Gauge: 10
EN 388 (X32X), EN 407 (X1XXXX); Cat. II
Sertifiseringer: EN 388 (X32X), EN 407 (X1XXXX); Cat. II