Vacuum pump, manual, with gauge
181-0256EA 500 NOK
EDSCP03078
Vacuum pump, manual, with gauge
Vacuum pump with a pressure gauge for the experiments with vacuum. Suitable for eg evacuation of air Magdeburg hemispheres.