Avtrekkskap, for skole
Leverandør: EDUCATIONSCIENCE COLLECTION P
765-0262EA 0 NOK
765-0262
Avtrekkskap, for skole
Avtrekksskap og kabinetter Avtrekksskap til apotek
School drawer SÖ.
Fume cupboard of simple model. White lacquered aluminum profiles and plexiglass drawbar. Stainless steel work surface. Base cabinet with sliding doors. Cold water tap, gas pipe, grounded electrical panel with 3 sockets and switch for lighting. Mandatory fuse box alarm will be added. Socket diam 160 mm and electrical connection on the roof.
School drawer Demo SÖ mobile
The same fume cupboard but mobile and the sides consisting of plexiglass. Slide-bearing rubber wheels.