Norway
Order Entry
Human thrombin (from plasma)
Human thrombin (from plasma)
  605195-5000
 :  
Human thrombin (from plasma)
  605195-5000
 :  
 : 605195-5000
 : CALB605195-5000
kr 36 475,00
  1 * 5.000 EE
  • Enzyme name:
    Thrombin
  • Enzyme activity:
    ≥2800 NIH units/mg
  • Cat. no.:
    605195-5000
Designed for use in thrombin clotting time tests. Key enzyme that activates factor XIII and converts fibrinogen into fibrin.