Ditizon Reag. Ph. Eur. analytisk reagens

Warning


M-Clarity™ quality level = MQ200

1.03092.0025EA 2080 NOK
1.03092.0025 1.03092.0005
Ditizon Reag. Ph. Eur. analytisk reagens
Ditizon
Formel: C₆H₅NHNHCSN=NC₆H₅
MW: 256,33 g/mol
Kokepunkt: 406 °C (1013 hPa)
Smeltepunkt: 168 °C
MDL nummer: MFCD00003025
CAS-nummer: 60-10-6
EINECS: 200-454-1
Merck Indeks: 13,03413

Bestill nå

Spesifikasjon Testresultater

Melting Point 168 °C (decomposition)
Molar Mass 256.32 g/mol
Bulk Density 250 kg/m3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR