Indole-3-carbaldehyd, Sigma-Aldrich®
Leverandør: Merck
M-Clarity™ quality level = MQ200
8.04726.0025EA
925
NOK
8.04726.0025
8.04726.9999
Indole-3-carbaldehyd, Sigma-Aldrich®
Indole-3-carbaldehyd
Formel:
C₉H₇NO MW: 145,16 g/mol Smeltepunkt: 192…195 °C Tetthet: 1,34 g/cm³ (20 °C) |
MDL nummer:
MFCD00005622 CAS-nummer: 487-89-8 EINECS: 207-665-8 |
Spesifikasjon Testresultater
Solubility | (20 °C) slightly soluble |
Melting Point | 192 - 195 °C |
Molar Mass | 145.16 g/mol |
Bulk Density | 200 - 300 kg/m3 |
Flash Point | 240 °C |
Ignition Point |
|
Synonyms | Indol-3-aldehyde |
Hill Formula | C9H7NO |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...