5-Nitrobenzimidazole
Leverandør: Molekula
MOLE28330984-100GEA
0
NOK
MOLE28330984-100G
5-Nitrobenzimidazole
5-Nitrobenzimidazole
Formel:
C₇H₅N₃O₂ MW: 163,14 g/mol Smeltepunkt: 200…203 °C |
MDL nummer:
MFCD00005604 CAS-nummer: 94-52-0 EINECS: 202-341-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...