Poles, 2 pcs
Leverandør: Mekoflex
765-1078EA 1075 NOK
MEKF194765-828
Poles, 2 pcs
Two identical, laminated pole shoes made of transformer sheet metal. One side 75×40 mm, flat ground and metal glossy. Other pages painted.
  • Dimensions: cross section 40 × 40 mm, length 75 mm