Norway
Order Entry
Lens with variable focal length
Lens with variable focal length
  763-0325
undefined
Lens with variable focal length
  763-0325
  1 * 1 Items
  • Description:
    Lens with variable focal length
  • Pk:
    1
Lens made of silicone rubber, mounted in a screen. The shape can be varied steplessly from convex to concave by changing the water volume in the lens. If an object is placed in front of the lens, the object can be projected on a posterior screen (retina). The focus is adjusted with the volume of water in the lens.