Poster, Spectrum
Leverandør: EDUCATIONSCIENCE COLLECTION P
766-0260EA 800 NOK
766-0260
Poster, Spectrum
Tabeller
Reproduces in multicolor printing a number of different absorption and emission spectra including solar spectrum with the main Fraunhofer lines and the continuous spectrum from a black body at 6000 K. Wavelength scale in nanometers below each spectrum.
  • Size 74 × 107 cm