Norway
Order Entry
Rotors for Centrifuge 5430/5430 R
Rotors for Centrifuge 5430/5430 R
Supplier:  EPPENDORF
   Sustainable Options Available
Certificates
undefined
Rotors for Centrifuge 5430/5430 R
Supplier:  EPPENDORF
   Sustainable Options Available
Specifications
  • Catalog No:
  • 521-1740
  • 521-1940
  • 521-2067
  • 521-2039
  • 521-1741
  • 521-2016
  • 521-2017
  • 521-2013
  • 521-2068
  • 521-2008
  • 521-1941
  • 521-2010
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Specifications
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • Swing out rotor A-2-MTP with 2 buckets and aluminium lid, wind shield, 2 MTP, PCR, and DWP (max. 0,5 ml), 2204×g, 4680 min⁻¹
    Swing out rotor A-2-MTP with 2 buckets and aluminium lid, wind shield, 2 MTP, PCR, and DWP (max. 0,5 ml), 2204×g, 4680 min⁻¹
    521-2010
    5427700.005
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 18 000,00
    kr 18 000,00
    1 * 1 Items
       kr 18 000,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
    Environmental certification
  • Fixed angle rotor F-45-64-5-PCR, 64×0,2 ml PCR tubes, with aluminium lid and adapters, 13543×g, 11800 min⁻¹
    Fixed angle rotor F-45-64-5-PCR, 64×0,2 ml PCR tubes, with aluminium lid and adapters, 13543×g, 11800 min⁻¹
    521-2016
    5427714.006
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 10 925,00
    kr 10 925,00
    1 * 1 Items
       kr 10 925,00
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Fixed angle rotor F-45-48-11, 48×1,5/2,0 ml, with rotor lid, 18210×g, 12700 min⁻¹
    Fixed angle rotor F-45-48-11, 48×1,5/2,0 ml, with rotor lid, 18210×g, 12700 min⁻¹
    521-2067
    5427755.004
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 7 550,00
    kr 7 550,00
    1 * 1 Items
       kr 7 550,00
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Fixed angle rotor F-45-30-11, PTFE coated, includes rotor lid, 30×1,5/2,0 ml tubes, 20817×g, 14000 min⁻¹
    Fixed angle rotor F-45-30-11, PTFE coated, includes rotor lid, 30×1,5/2,0 ml tubes, 20817×g, 14000 min⁻¹
    521-2008
    5427712.003
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 6 625,00
    kr 6 625,00
    1 * 1 Items
       kr 6 625,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
    Environmental certification
  • Fixed angle rotor F-45-18-17-Cryo, 18 cryotubes, with PP lid and adapters, 8324×g, 11800 min⁻¹
    Fixed angle rotor F-45-18-17-Cryo, 18 cryotubes, with PP lid and adapters, 8324×g, 11800 min⁻¹
    521-2013
    5427705.007
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 6 950,00
    kr 6 950,00
    1 * 1 Items
       kr 6 950,00
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Fixed angle rotor F-35-6-30, 6×15 ml/50 ml Falcon tubes or 12×Vacutainer® tubes, blood collection tubes, Oak ridge tubes, with aluminium lid and adapter, 7745×g, 7830 min⁻¹
    Fixed angle rotor F-35-6-30, 6×15 ml/50 ml Falcon tubes or 12×Vacutainer® tubes, blood collection tubes, Oak ridge tubes, with aluminium lid and adapter, 7745×g, 7830 min⁻¹
    521-2017
    5427716.009
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 18 000,00
    kr 18 000,00
    1 * 1 Items
       kr 18 000,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
    Environmental certification
  • Aerosol-tight fixed angle rotor FA-45-48-11, 48×1,5/2,0 ml, with rotor lid, 18210×g, 12700 min⁻¹
    Aerosol-tight fixed angle rotor FA-45-48-11, 48×1,5/2,0 ml, with rotor lid, 18210×g, 12700 min⁻¹
    521-1940
    5427754.008
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 10 400,00
    kr 10 400,00
    1 * 1 Items
       kr 10 400,00
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Aerosol-tight fixed angle rotor FA-45-30-11, 30×1,5/2,0 ml, with aluminium lid and PTFE coating, 20817×g, 14000 min⁻¹
    Aerosol-tight fixed angle rotor FA-45-30-11, 30×1,5/2,0 ml, with aluminium lid and PTFE coating, 20817×g, 14000 min⁻¹
    521-1741
    5427753.001
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 8 925,00
    kr 8 925,00
    1 * 1 Items
       kr 8 925,00
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Aerosol-tight fixed angle rotor FA-45-24-11-HS, 24×1,5/2,0 ml, with aluminium lid and PTFE coating, 30130×g, 17500 min⁻¹
    Aerosol-tight fixed angle rotor FA-45-24-11-HS, 24×1,5/2,0 ml, with aluminium lid and PTFE coating, 30130×g, 17500 min⁻¹
    521-2039
    5427710.000
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 9 250,00
    kr 9 250,00
    1 * 1 Items
       kr 9 250,00
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Aerosol-tight fixed angle rotor FA-45-24-11-Kit, 24 spin columns, with aluminium lid, 19090×g, 13200 min⁻¹
    Aerosol-tight fixed angle rotor FA-45-24-11-Kit, 24 spin columns, with aluminium lid, 19090×g, 13200 min⁻¹
    521-1740
    5427752.005
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 11 450,00
    kr 11 450,00
    1 * 1 Items
       kr 11 450,00
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Aerosol-tight fixed angle rotor FA-45-16-17, for 16×5,0 ml Eppendorf Tubes®, with lid, 21191×g, 14200 min⁻¹
    Aerosol-tight fixed angle rotor FA-45-16-17, for 16×5,0 ml Eppendorf Tubes®, with lid, 21191×g, 14200 min⁻¹
    521-1941
    5427750.002
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 11 975,00
    kr 11 975,00
    1 * 1 Items
       kr 11 975,00
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Aerosol-tight swing-out bucket rotor S-24-11-AT, 24×1,5/2,0 ml, with rotor lid, 16049×g, 12700 min⁻¹
    Aerosol-tight swing-out bucket rotor S-24-11-AT, 24×1,5/2,0 ml, with rotor lid, 16049×g, 12700 min⁻¹
    521-2068
    5427757.007
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 16 350,00
    kr 16 350,00
    1 * 1 Items
       kr 16 350,00
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
Adapters for F-45-30-11, FA-45-48-11, FA-45-24-11, FA-45-24-11-HS Kits
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Price
    Availability
    Price
  • 09
    Adapter, for 1 PCR tube (0,2 ml, max, Ø 6 mm), for all 1,5/2,0 ml rotors, 6 pieces
    6
    9,40
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - kr 210,00
    kr 210,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Price
    Availability
    Price
  • 08
    Adapter, for 1 micro test tube (0,4 ml, max, Ø 6 mm), 6 pieces
    6
    9,40
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - kr 220,00
    kr 220,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Price
    Availability
    Price
  • 04
    Adapter for 0,5 ml microcentrifuge tube
    6
    9,40
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - kr 210,00
    kr 210,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 03
    Rotor lid FA-45-30-11, aerosol-tight, for centrifuge 5430/5430R
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 275,00
    kr 3 275,00
Adapters for F-35-6-30
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 20
    Adapter, for 1×5.0 ml Eppendorf tubes®, F-35-6-30 large bore, 2 pieces
    2
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 300,00
    kr 1 300,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 19
    Adapter, for 1×5.0 ml Eppendorf tubes®, F-35-6-30 small bore, 2 pieces
    2
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 300,00
    kr 1 300,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 15
    Adapter, for 1 tubes 65 - 89 mm, for rotor F-35-6-30, small rotor bore, 2 pieces
    2
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 300,00
    kr 1 300,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 17
    Adapter, for 1 tubes 65 - 89 mm, for rotor F-35-6-30, large rotor bore, 2 pieces
    2
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 300,00
    kr 1 300,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 16
    Adapter, for 1 tubes 90 - 110 mm, for rotor F-35-6-30, small rotor bore, 2 pieces
    2
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 300,00
    kr 1 300,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Price
    Availability
    Price
  • 11
    Adapter 17 mm RND, for small bore 2 packs (5430)
    2
    58,70
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 325,00
    kr 1 325,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Price
    Availability
    Price
  • 10
    Adapter 16 mm RND, for large bore, 2 packs (5430)
    2
    58,70
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 325,00
    kr 1 325,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Price
    Availability
    Price
  • 14
    Adapter 17 mm RND, for large bore, 2 packs (5430)
    2
    58,70
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 325,00
    kr 1 325,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 18
    Adapter, for 1 tubes 90 - 110 mm, for rotor F-35-6-30, large rotor bore, 2 pieces
    2
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 300,00
    kr 1 300,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Price
    Availability
    Price
  • 08
    Adapter 16 mm RND, for small bore 2 packs (5430)
    2
    58,70
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 325,00
    kr 1 325,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Price
    Availability
    Price
  • 05
    Adapter, for 1 conical tubes 15 ml, for rotor F-35-6-30, small rotor bore, 2 pieces
    2
    53,70
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 200,00
    kr 1 200,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Price
    Availability
    Price
  • 12
    Adapter, for 1 round bottom tubes 30 ml, F-35-6-30 large bore, 2 pieces
    2
    47,70
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 075,00
    kr 1 075,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Price
    Availability
    Price
  • 13
    Adapter, for 1 round bottom tubes 50 ml, F-35-6-30 large bore, 2 pieces
    2
    47,70
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 075,00
    kr 1 075,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Price
    Availability
    Price
  • 06
    Adapter, for 1 conical tubes 50 ml, for rotor F-35-6-30, large rotor bore, 2 pieces
    2
    31,90
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 710,00
    kr 710,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Price
    Availability
    Price
  • 04
    Adapter, for 1 Centriprep®, F-35-6-30 large bore, 6 pieces
    6
    87,30
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - kr 1 950,00
    kr 1 950,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Price
    Availability
    Price
  • 09
    Adapter 13 mm RND, for large bore, 2 packs (5430)
    2
    58,70
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 325,00
    kr 1 325,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Price
    Availability
    Price
  • 07
    Adapter, for 1 round bottom tubes 2.6-7ml, F-35-6-30 small bore, 2 pieces
    2
    58,70
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 325,00
    kr 1 325,00
Adapters for F-45-18-17-Cryo
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 22
    Adapter, for 1 HPLC vessel 1.5 ml, for rotor F-45-18-17-Cryo, 18 pieces
    18
    Log in for availability
    Log in for availability
    1 * 18 Items
    1 * 18 Items - kr 1 850,00
    kr 1 850,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 24
    Adapter Rd 13 mm verpackt (6×)
    6
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - kr 980,00
    kr 980,00
Adapters for A-2-MTP
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 50
    Adapter, for 1 PCR plate (96 wells), 2 pieces
    2
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 2 150,00
    kr 2 150,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 51
    Adapter, for 1 PCR plate (384 wells), 2 pieces
    2
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 725,00
    kr 1 725,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 134
    SBS adapter, for 1 plate with frame in SBS format, 2 pieces
    2
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 975,00
    kr 1 975,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 35
    MTP bucket for rotor A-2-MTP, 2 pieces
    2
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 6 800,00
    kr 6 800,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 34
    CombiSlide adapter, for 12 slides combi slide, 2 pieces
    2
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 4 450,00
    kr 4 450,00
Adapters for F-45-64-5-PCR
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Price
    Availability
    Price
  • 23
    Adapter, for 1 PCR strip, for rotor F-45-64-5-PCR, 4 pieces
    4
    110,00
    Log in for availability
    Log in for availability
    1 * 4 Items
    1 * 4 Items - kr 2 450,00
    kr 2 450,00
Adapters for FA-45-16-17
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 129
    Adapter, for 1 vessel 1.5 - 2.0 ml, for all 5.0 ml rotors, 4 pieces
    4
    Log in for availability
    Log in for availability
    1 * 4 Items
    1 * 4 Items - kr 520,00
    kr 520,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 128
    Adapter, for 1 cryo tubes, for all 5.0 ml rotors, 4 pieces
    4
    Log in for availability
    Log in for availability
    1 * 4 Items
    1 * 4 Items - kr 520,00
    kr 520,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 127
    Adapter, for 1 HPLC tubes, for all 5.0 ml rotors, 4 pieces
    4
    Log in for availability
    Log in for availability
    1 * 4 Items
    1 * 4 Items - kr 520,00
    kr 520,00
Adapters for S-24-11-AT
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 01
    Tubes holder for S-24-11-AT, for 4×1.5 ml/2.0 ml Eppendorf tubes, 2 pieces
    2
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - kr 1 275,00
    kr 1 275,00
Spare Rotor Lids
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 36
    Rotor lid for A-2-MTP, aluminum, for centrifuge 5430/5430R
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 4 450,00
    kr 4 450,00
Product Information
  • Catalog #
    Description
    Pk
    Availability
    Price
  • Rotor lid for F-45-64-5-PCR, aluminum, for centrifuge 5430/5430R
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 200,00
    kr 2 200,00
Product Information
  • Catalog #
    Description
    Pk
    Availability
    Price
  • Rotor lid for F-45-18-17-Cryo, polypropylene, for centrifuge 5430/5430R
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 075,00
    kr 2 075,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 22
    Rotor lid for F-35-6-30, aluminum, for centrifuge 5430/5430R
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 075,00
    kr 3 075,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 28
    Rotor lid FA-45-24-11-Kit, aerosol-tight, for centrifuge 5430/5430R
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 850,00
    kr 2 850,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 29
    Rotor lid FA-45-30-11, for centrifuge 5430/5430R
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 075,00
    kr 2 075,00
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 02
    Rotor lid FA-45-24-11-HS, aerosol-tight, for centrifuge 5430/5430R
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 275,00
    kr 3 275,00