Wet/Dry Vacuum Cleaner, Delfin LC1100WD
Catalog # TEKMHWVC-IDVCPWD
Supplier: TEKNOMEK
Specifications
- Description:Vaccum cleaner (wet and dry), 430×430×730 mm
- Pk:1
Specifications
The LC1100 WD is an industrial vacuum for clean rooms, ideal for removing liquids and sanitising cleanrooms.
- Autoclavable (header excluded)
- Single phase throughflow engine
- No dirt traps
- Up to 4 stages of filtration
- 304 stainless steel construction, with 316 stainless steel for components in contact with material
- 5 Litre capacity
- UK plug included
Multiple levels of filtration (HEPA and ULPA available) ensure that the air released by the vacuum cleaner is pure, while smooth surfaces and stainless steel construction make cleaning, maintenance and sanitisation easier.
The vacuum cleaner and its accessories (motor head excluded) are autoclavable for complete and safe sanitising of the vacuum cleaner.