Caps, KIM-KAP™, disposable, KIMBLE®
PP, coloured.
These caps are designed to fit culture tubes with outer Ø ranging from 13 to 38 mm.
- For Ø ext.ColourPk
- 391-0495SP73660-1313 mmNatural1001 * 100 Itemskr 430,00
Specifications
Environmentally Preferable - 391-0501SP73662-1313 mmRed1001 * 100 Itemskr 450,00
Specifications
Environmentally Preferable - 391-0509SP73663-1313 mmGreen1001 * 100 Itemskr 465,00
Specifications
Environmentally Preferable - 391-0514SP73664-1313 mmYellow1001 * 100 Itemskr 495,00
Specifications
Environmentally Preferable - 391-0519SP73665-1313 mmBlue1001 * 100 Itemskr 405,00
Specifications
Environmentally Preferable - 391-0496SP73660-1616 mmNatural1001 * 100 Itemskr 495,00
Specifications
Environmentally Preferable - 391-0503SP73662-1616 mmRed1001 * 100 Itemskr 495,00
Specifications
Environmentally Preferable - 391-0510SP73663-1616 mmGreen1001 * 100 Itemskr 495,00
Specifications
Environmentally Preferable - 391-0515SP73664-1616 mmYellow1001 * 100 Itemskr 495,00
Specifications
Environmentally Preferable - 391-0520SP73665-1616 mmBlue1001 * 100 Itemskr 495,00
Specifications
Environmentally Preferable - 391-0497SP73660-1818 mmNatural1001 * 100 Itemskr 720,00
Specifications
Environmentally Preferable - 391-0505SP73662-1818 mmRed1001 * 100 Itemskr 515,00
Specifications
Environmentally Preferable - 391-0511SP73663-1818 mmGreen1001 * 100 Itemskr 580,00
Specifications
Environmentally Preferable - 391-0516SP73664-1818 mmYellow1001 * 100 Itemskr 580,00
Specifications
Environmentally Preferable - 391-0521SP73665-1818 mmBlue1001 * 100 Itemskr 805,00
Specifications
Environmentally Preferable - 391-0498SP73660-2020 mmNatural5001 * 500 Itemskr 3 050,00
Specifications
Environmentally Preferable - 391-0507SP73662-2020 mmRed1001 * 100 Itemskr 805,00
Specifications
Environmentally Preferable - 391-0512SP73663-2020 mmGreen1001 * 100 Itemskr 1 300,00
Specifications
Environmentally Preferable - 391-0517SP73664-2020 mmYellow1001 * 100 Itemskr 1 300,00
Specifications
Environmentally Preferable - 391-0523SP73665-2020 mmBlue1001 * 100 Itemskr 1 300,00
Specifications
Environmentally Preferable - 391-0499SP73660-2525 mmNatural5001 * 500 Itemskr 4 175,00
Specifications
Environmentally Preferable - 391-0508SP73662-2525 mmRed1001 * 100 Itemskr 1 575,00
Specifications
Environmentally Preferable - 391-0513SP73663-2525 mmGreen1001 * 100 Itemskr 1 575,00
Specifications
Environmentally Preferable - 391-0518SP73664-2525 mmYellow1001 * 100 Itemskr 1 575,00
Specifications
Environmentally Preferable - 391-0500SP73660-3838 mmNatural501 * 50 Itemskr 1 025,00
Specifications
Environmentally Preferable