Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
kr 0,00
Order Entry
Pyridinium dichromate
MOLEM41679983
:
Molekula
:
Pyridinium dichromate
MOLEM41679983
:
Molekula
:
: M41679983
: MOLEM41679983
kr 2 475,00
1 * 25 g
-
+
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
532.19 g/mol
:
152…153 °C
:
243-478-8
:
3087
:
5.1,II
:
20039-37-6
:
C₁₀N₁₂Cr₂N₂O₇
:
MFCD00013105