Anti-ATP2B2 Rabbit Polyclonal Antibody
PRSI22-523
New Product
- Antibody type:Primary
- Antigen name:ATPase plasma membrane Ca2+ transporting 2
- Antigen symbol:ATP2B2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:491
- Antigen synonyms:Ca++ transporting|plasma membrane Ca2+ pump 2|PMCA2a|plasma membrane calcium ATPase|plasma membrane calcium-transporting ATPase 2|plasma membrane 2|plasma membrane calcium pump|PMCA2i|PMCA2|plasma membrane Ca(2+)-ATPase|ATPase
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human ATP2B2 (NP_001674.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: ATP2B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat