Norway
Order Entry
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ UV375)
 
undefined
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ UV375)

 

  • Catalog No:
  • AATB100330X0
  • AATB100330X1
  • Antigensymbol:
  • CD3e
  • CD3e
  • Antigen-navn:
  • CD3e antigen
  • CD3e antigen
  • Antistofftype:
  • Primary
  • Primary
  • Klonalitet:
  • Monoclonal
  • Monoclonal
  • Konjugering:
  • mFluor™ UV375
  • mFluor™ UV375
  • Klon:
  • SK7
  • SK7
  • Reaktivitet:
  • Human
  • Human
  • Vert:
  • Mouse
  • Mouse
  • ImmunoFluorescens:
  • Yes
  • Yes
  • Flowcytometri:
  • Yes
  • Yes
  • Rensing:
  • Affinity chromatography
  • Affinity chromatography
  • Kyssabsorpsjon:
  • No
  • No
  • Lagringstemp:
  • 2...8 °C with minimised light exposure, do not freeze
  • 2...8 °C with minimised light exposure, do not freeze
  • Frakttemp.:
  • 20
  • 20

 

 

  •  
     
    Pakketype
    Fp
     
     
  • AATB100330X0
    100330X0
    Vial
    100 Tests
       
       
    1 * 1 Each
    1 * 1 Each - kr 4 350,00
    kr 4 350,00
    1 * 1 Each
       kr 4 350,00

     
     
    Specifications
    Antigensymbol
    CD3e
    Antigen-navn
    CD3e antigen
    Environmentally Preferable
    Antistofftype
    Primary
    Klonalitet
    Monoclonal
    Konjugering
    mFluor™ UV375
    Klon
    SK7
    Reaktivitet
    Human
    Vert
    Mouse
    Isotype
    ImmunoFluorescens
    Yes
    Flowcytometri
    Yes
    Rensing
    Affinity chromatography
    Kyssabsorpsjon
    No
    Lagringstemp
    2...8 °C with minimised light exposure, do not freeze
    Frakttemp.
    20
  • AATB100330X1
    100330X1
    Vial
    500 Tests
       
       
    1 * 1 Each
    1 * 1 Each - kr 16 275,00
    kr 16 275,00
    1 * 1 Each
       kr 16 275,00

     
     
    Specifications
    Antigensymbol
    CD3e
    Antigen-navn
    CD3e antigen
    Environmentally Preferable
    Antistofftype
    Primary
    Klonalitet
    Monoclonal
    Konjugering
    mFluor™ UV375
    Klon
    SK7
    Reaktivitet
    Human
    Vert
    Mouse
    Isotype
    ImmunoFluorescens
    Yes
    Flowcytometri
    Yes
    Rensing
    Affinity chromatography
    Kyssabsorpsjon
    No
    Lagringstemp
    2...8 °C with minimised light exposure, do not freeze
    Frakttemp.
    20