Norway
Order Entry
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PE/iFluor® 700)
undefined
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PE/iFluor® 700)
  • Catalog No:
  • AATB100321X0
  • AATB100321X1
  • AATB100321X2
  • Antigen symbol:
  • CD3e
  • CD3e
  • CD3e
  • Antigen name:
  • CD3e antigen
  • CD3e antigen
  • CD3e antigen
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)/iFluor® 700
  • PE (Phycoerythrin)/iFluor® 700
  • PE (Phycoerythrin)/iFluor® 700
  • Clone:
  • UCHT1
  • UCHT1
  • UCHT1
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Affinity chromatography
  • Cross adsorption:
  • No
  • No
  • No
  • Storage temperature:
  • 2...8 °C with minimised light exposure, do not freeze
  • 2...8 °C with minimised light exposure, do not freeze
  • 2...8 °C with minimised light exposure, do not freeze
  • Shipping temperature:
  • 20
  • 20
  • 20
  •  
    Pack type
    Pk
  • AATB100321X0
    100321X0
    Vial
    25 Tests
    1 * 1 Each
    1 * 1 Each - kr 3 750,00
    kr 3 750,00
    1 * 1 Each
       kr 3 750,00
    Specifications
    Antigen symbol
    CD3e
    Antigen name
    CD3e antigen
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    PE (Phycoerythrin)/iFluor® 700
    Clone
    UCHT1
    Reactivity
    Human
    Host
    Mouse
    Isotype
    ImmunoFluorescence
    Yes
    Flow cytometry
    Yes
    Purification
    Affinity chromatography
    Cross adsorption
    No
    Storage temperature
    2...8 °C with minimised light exposure, do not freeze
    Shipping temperature
    20
  • AATB100321X1
    100321X1
    Vial
    100 Tests
    1 * 1 Each
    1 * 1 Each - kr 10 400,00
    kr 10 400,00
    1 * 1 Each
       kr 10 400,00
    Specifications
    Antigen symbol
    CD3e
    Antigen name
    CD3e antigen
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    PE (Phycoerythrin)/iFluor® 700
    Clone
    UCHT1
    Reactivity
    Human
    Host
    Mouse
    Isotype
    ImmunoFluorescence
    Yes
    Flow cytometry
    Yes
    Purification
    Affinity chromatography
    Cross adsorption
    No
    Storage temperature
    2...8 °C with minimised light exposure, do not freeze
    Shipping temperature
    20
  • AATB100321X2
    100321X2
    Vial
    500 Tests
    1 * 1 Each
    1 * 1 Each - kr 35 225,00
    kr 35 225,00
    1 * 1 Each
       kr 35 225,00
    Specifications
    Antigen symbol
    CD3e
    Antigen name
    CD3e antigen
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    PE (Phycoerythrin)/iFluor® 700
    Clone
    UCHT1
    Reactivity
    Human
    Host
    Mouse
    Isotype
    ImmunoFluorescence
    Yes
    Flow cytometry
    Yes
    Purification
    Affinity chromatography
    Cross adsorption
    No
    Storage temperature
    2...8 °C with minimised light exposure, do not freeze
    Shipping temperature
    20