Anti-CD9 Mouse Monoclonal Antibody [clone: HI9a]
STMC100-0138
- Antigen name:CD9
- Clonality:Monoclonal
- Clone:HI9a
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Baboon,Capuchin monkey,African green monkey,Cynomolgus monkey,Rhesus monkey,Chimpanzee,Human
- Gene ID:928
- Pk:100 µG