/
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Mouse IgG ELISA Antibody Pair Kit
STMC01998
:
STEMCELL Technologies
:
Mouse IgG ELISA Antibody Pair Kit
STMC01998
:
STEMCELL Technologies
:
: 01998
: STMC01998
1 * 1 Each
-
+
Pk:
1 KIT
Primary antibody reactivity:
Mouse
Target protein:
IgG