Norway
Order Entry
Anti-IgG Goat Polyclonal Antibody (iFluor® 820)
New Product
 
Anti-IgG Goat Polyclonal Antibody (iFluor® 820)
New Product

 

  • Catalog No:
  • AATB48058
  • AATB48059
  • Antigen symbol:
  • IgG
  • IgG
  • Antigen name:
  • Immunoglobulin G
  • Immunoglobulin G
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • iFluor® 820
  • iFluor® 820
  • Reactivity:
  • Rabbit
  • Rabbit
  • Host:
  • Goat
  • Goat
  • Isotype:
  • IgG (H, L)
  • IgG (H, L)
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • Freeze (<−15 °C); Minimise light exposure
  • Freeze (<−15 °C); Minimise light exposure
  • Shipping temperature:
  • 20
  • 20

 

 

  •  
     
    Pack type
    Pk
     
     
  • AATB48058
    48058
    Vial
    200 µG
       
       
    1 * 1 Each
    1 * 1 Each - kr 1 625,00
    kr 1 625,00
    1 * 1 Each
       kr 1 625,00

     : 48058
     : AATB48058
     
    New Product
     
    Specifications
    Antigen symbol
    IgG
    Antigen name
    Immunoglobulin G
    Environmentally Preferable
    Antibody type
    Secondary
    Clonality
    Polyclonal
    Conjugation
    iFluor® 820
    Reactivity
    Rabbit
    Host
    Goat
    Isotype
    IgG (H, L)
    Purification
    Affinity chromatography
    Cross adsorption
    No
    Storage temperature
    Freeze (<−15 °C); Minimise light exposure
    Shipping temperature
    20
    New Product
  • AATB48059
    48059
    Vial
    1 mg
       
       
    1 * 1 mg
    1 * 1 mg - kr 3 000,00
    kr 3 000,00
    1 * 1 mg
       kr 3 000,00

     : 48059
     : AATB48059
     
    New Product
     
    Specifications
    Antigen symbol
    IgG
    Antigen name
    Immunoglobulin G
    Environmentally Preferable
    Antibody type
    Secondary
    Clonality
    Polyclonal
    Conjugation
    iFluor® 820
    Reactivity
    Rabbit
    Host
    Goat
    Isotype
    IgG (H, L)
    Purification
    Affinity chromatography
    Cross adsorption
    No
    Storage temperature
    Freeze (<−15 °C); Minimise light exposure
    Shipping temperature
    20
    New Product