Supelco® TraceCERT® Organic Reference Standard, Ethiofencarb-sulfone
CRM05139-10MG
: Merck
:
- Pk:10 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:53380-23-7
- Description:Ethiofencarb-sulfone
- Storage conditions:2…8 °C
- Hazard class description:H302
- Shipping conditions:WICE
- MDL No.:MFCD01311808
- M-Clarity quality level:MQ300
- Molecular weight:257,31