Norway
Order Entry
Male-Female Luer Lock Caps
  BDAMTP-MF-O
Male-Female Luer Lock Caps
  BDAMTP-MF-O
kr 770,00
  1 * 100 Items
  • Colour:
    Orange
  • Description:
    Male-Female Luer lock caps, sterile
  • Pk:
    100
Caps for Luer lock and Luer slip. Combistoppers for female and male Luer lock.
  • Individual blisters (except for 394080 and 302834 that come in a tray)
DEHP or natural rubber latex are not part of the material formulation.