
Biphenylene 99+%
ACRO279742500
:
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
- Appearance:Yellow Needles
- Infrared spectrum:Conforms
- Melting point:109 to 112 °C
- GC:≥99.0 %
:
:152.2 g/mol
:262 °C (1013 hPa)
:109…112 °C
:1,23 g/cm³ (25 °C)
:Ambient
:3077
:9,III
:GHS09
:259-79-0
:C₁₂H₈
:MFCD00001110