Urine and biological specimen containers, with screw caps
: KARTELL
PS, transparent with PE or PP screw caps.
- 60 ml with PE screw caps and 200 ml with PP screw caps
- Quick opening and closing twist cap, leakproof
Ideal for collecting, storing and transporting urine samples and biological specimens.
* Aseptic containers individually wrapped, with label.
- CapacityExt.ØHeightPk
- 216-0264563060 ml39 mm65 mm6001 * 600 Itemskr 5 925,00: 5630: KART5630
Specifications
Environmentally Preferable - 216-0442563160* ml39 mm65 mm4001 * 400 Itemskr 5 050,00: 5631: KART5631
Specifications
Environmentally Preferable - 2596200 ml66 mm85,5 mm5001 * 500 Itemskr 8 750,00: 2596: KART2596
Specifications
Environmentally Preferable - 2696200* ml66 mm85,5 mm2001 * 200 Itemskr 4 525,00: 2696: KART2696
Specifications
Environmentally Preferable