Norway
Order Entry
Desiccator cabinets, SICCO Maxi series
 :  Bohlender
undefined
Desiccator cabinets, SICCO Maxi series
 :  Bohlender
  •  
    Shelves
    With
    W×D×H ext.
    W×D×H int.
    Pk
  • Maxi 1 for drying/storage (280 L)
    Maxi 1 for drying/storage (280 L)
    467-0044
    V1852-01
    4
    1 chamber and 1 door
    560×580×1150 mm
    495×540×1030 mm
    1
    1 * 1 Items
    1 * 1 Items - kr 72 675,00
    kr 72 675,00
    1 * 1 Items
       kr 72 675,00
    Specifications
    Environmentally Preferable
  • Maxi 2 for drying/storage (91 L)*
    Maxi 2 for drying/storage (91 L)*
    467-0045
    V1853-01
    4
    2 chambers and 2 doors
    560×580×1150 mm
    495×540×500* mm
    1
    1 * 1 Items
    1 * 1 Items - kr 91 425,00
    kr 91 425,00
    1 * 1 Items
       kr 91 425,00
    Specifications
    Environmentally Preferable