Petri dishes
Made from crystal clear PS for best possible use in microscopic applications. Versions with or without vents.
- Easily stackable
- Heavy construction for good thermal resistance when working with hot agar
- With vents for better gas exchange or without vents for long incubation times
- Catalog No:
- 391-0488
- 391-0491
- 391-0558
- 391-0486
- 391-3665
- 391-0487
- 391-0261
- 391-2079
- 391-3661
- 391-0493
- 391-3660
- 391-3655
- 391-0490
- 391-2072
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- VersionØHeightSterilityPackedPk
- 391-2072627102_740With 3 vents35 mm10 mmNon sterile10/sleeve7401 * 740 Itemskr 3 100,00
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 391-0486627161_740With 3 vents35 mm10 mmSterile10/sleeve7401 * 740 Itemskr 2 750,00
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 391-3655628102_600With 6 vents60 mm15 mmNon sterile20/sleeve6001 * 600 Itemskr 1 775,00
Specifications
Environmentally PreferableEnvironmental certification - 628161_600With 3 vents60 mm15 mmSterile20/sleeve6001 * 600 Itemskr 2 125,00
Specifications
Environmentally PreferableEnvironmental certification - 391-3661632180_480Without vents90 mm16 mmNon sterile20/sleeve4801 * 480 Itemskr 2 000,00
Specifications
Environmentally PreferableEnvironmental certification - 391-0261635102_480With 2 compartments (20 ml)90 mm16 mmNon sterile20/sleeve4801 * 480 Itemskr 1 900,00
Specifications
Environmentally PreferableEnvironmental certification - 391-2079664102_360With 3 vents92 mm20 mmNon sterile15/sleeve3601 * 360 Itemskr 1 175,00
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 664161_360With 3 vents92 mm20 mmSterile15/sleeve3601 * 360 Itemskr 1 375,00
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002) - 632181_480Without vents94 mm16 mmSterile20/sleeve4801 * 480 Itemskr 2 050,00
Specifications
Environmentally PreferableEnvironmental certification - 633180_480With 3 vents94 mm16 mmNon sterile20/sleeve4801 * 480 Itemskr 1 650,00
Specifications
Environmentally PreferableEnvironmental certification - 633181_480With 3 vents94 mm16 mmSterile20/sleeve4801 * 480 Itemskr 1 975,00
Specifications
Environmentally PreferableEnvironmental certification - 635161_480With 2 compartments (25 ml)94 mm15 mmSterile20/sleeve4801 * 480 Itemskr 2 350,00
Specifications
Environmentally PreferableEnvironmental certification - 391-3665639102_120With 3 vents145 mm20 mmNon sterile15/sleeve1201 * 120 Itemskr 1 050,00
Specifications
Environmentally PreferableEnvironmental certification - 639161_120With 3 vents, heavy design145 mm20 mmSterile15/sleeve1201 * 120 Itemskr 1 650,00
Specifications
Environmentally PreferableEnvironmental certification